Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-hydroxybutyrate 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
5405-41-4
C6H12O3
132.16
MFCD00004545
OMSUIQOIVADKIM-UHFFFAOYSA-N
ethyl 3-hydroxybutyrate,ethyl beta-hydroxybutyrate,butanoic acid, 3-hydroxy-, ethyl ester,butyric acid, 3-hydroxy-, ethyl ester,fema no. 3428,ethyl 3-hydroxybutyrate natural,ethyl dl-3-hydroxybutyrate,ethyl 1-3-hydroxybutyrate,dl-3-hydroxy-n-butyric acid ethyl ester,ethyl3-hydroxybutyrate
62572
CHEBI:87685
ethyl 3-hydroxybutanoate
CCOC(=O)CC(C)O
5405-41-4
C6H12O3
132.16
MFCD00004545
OMSUIQOIVADKIM-UHFFFAOYSA-N
ethyl 3-hydroxybutyrate,ethyl beta-hydroxybutyrate,butanoic acid, 3-hydroxy-, ethyl ester,butyric acid, 3-hydroxy-, ethyl ester,fema no. 3428,ethyl 3-hydroxybutyrate natural,ethyl dl-3-hydroxybutyrate,ethyl 1-3-hydroxybutyrate,dl-3-hydroxy-n-butyric acid ethyl ester,ethyl3-hydroxybutyrate
62572
CHEBI:87685
ethyl 3-hydroxybutanoate
CCOC(=O)CC(C)O
Boiling Point 170°C
Flash Point 64°C
Specific Gravity 1.01
Packaging Glass bottle
Density 1.0100g/mL
Refractive Index 1.4195 to 1.4215
Infrared Spectrum Authentic
Color Colorless
Quantity 25g
Assay Percent Range 99%
Linear Formula CH3CH(OH)CH2CO2C2H5
Beilstein 03, 309
Fieser 14,162
Solubility Information Solubility in water: soluble
Formula Weight 132.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 3-hydroxybutyrate, 99%
Show More Show Less

EINECSNumber : 226-456-2

TSCA : TSCA