Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%, Thermo Scientific™

Catalog No. AAH64533MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26510-52-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094022
InChI Key FQHXWZMJALFSJJ-UHFFFAOYSA-N
Synonym ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
PubChem CID 2736461
SMILES CCOC(=O)CC(=O)C1=CC=CC=N1

Specifications

Density 1.164 g/mL
Boiling Point 100°C to 104°C (0.3 mmHg)
Flash Point >200°C (392°F)
Refractive Index 1.516
Quantity 250 mg
Beilstein 145706
Sensitivity Light sensitive
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
Formula Weight 193.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.