Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-phenoxypropionate, 98%

Catalog No. aal0760609
Click to view available options
Quantity:
10 g
50 g

CAS: 22409-91-2 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22409-91-2
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026919
InChI Key TXKJOPXGYSFUNC-UHFFFAOYSA-N
PubChem CID 347973
IUPAC Name ethyl 3-phenoxypropanoate
SMILES CCOC(=O)CCOC1=CC=CC=C1

Specifications

Melting Point 22°C to 24°C
Density 1.084
Boiling Point 141°C to 143°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5005
Quantity 10 g
Beilstein 2331072
Synonym ethyl 3-phenoxypropionate, 3-phenoxypropanoic acid ethyl ester, acmc-20akz9, propanoic acid,3-phenoxy-, ethyl ester, propanoic acid, 3-phenoxy-, ethyl ester
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 3-phenoxypropionate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.