Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,4-trifluoro-2-methylacetoacetate, 96%

Catalog No. AAL1733503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1733503 1 g
AAL1733506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1733503 Supplier Thermo Scientific Chemicals Supplier No. L1733503
Add to Cart

CAS: 344-00-3 | C7H9F3O3 | 198.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-00-3
Molecular Formula C7H9F3O3
Molecular Weight (g/mol) 198.141
MDL Number MFCD00190635
InChI Key YLRGPBKEZVHOAW-UHFFFAOYSA-N
Synonym ethyl 2-methyl-4,4,4-trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-2-methyl-3-oxo-, ethyl ester, zlchem 125, acmc-20al0w, ethyl 2-methyltrifluoroacetoacetate, ethyl 2-methyl-4,4,4-trifluoro acetoacetate, ethyl alpha-methyl-4,4,4-trifluoroacetoacetate, ethyl4,4,4-trifluoro-2-methyl-3-oxobutanoate, ethyl 3-keto-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-2-methyl-3-oxo-butanoate
PubChem CID 2737204
IUPAC Name ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C(F)(F)F

Specifications

Density 1.258
Boiling Point 144°C to 146°C
Flash Point 47°C (116°F)
Refractive Index 1.37
Quantity 1 g
UN Number UN3272
Beilstein 1783620
Formula Weight 198.14
Percent Purity 96%
Chemical Name or Material Ethyl 4,4,4-trifluoro-2-methylacetoacetate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.