Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-aminophenylacetate, 98%, Thermo Scientific™

Catalog No. AC355160010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-70-0
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017569
InChI Key CFNDVXUTYPXOPG-UHFFFAOYSA-N
Synonym ethyl 4-aminophenylacetate, ethyl 2-4-aminophenyl acetate, p-aminophenylacetic acid ethyl ester, 4-aminophenylacetic acid ethyl ester, ethyl 4-aminophenyl acetate, ethyl p-aminophenylacetate, unii-5h8cfu7gnp, benzeneacetic acid, 4-amino-, ethyl ester, ethyl-4-aminophenylacetate
PubChem CID 225219
IUPAC Name ethyl 2-(4-aminophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)N

Specifications

Melting Point 48°C to 52°C
Color Beige to Pink-White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NHC6H4CHCOCHCH3
Quantity 1 g
Formula Weight 179.22
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Ethyl 4-aminophenylacetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.