Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-aminopiperidine-1-carboxylate, 98%

Catalog No. AAL1586614
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAL1586614

Supplier: Thermo Scientific Chemicals L1586614

CAS: 58859-46-4 | C8H16N2O2 | 172.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58859-46-4
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.228
MDL Number MFCD00006484
InChI Key GQQQULCEHJQUJT-UHFFFAOYSA-N
Synonym ethyl 4-amino-1-piperidinecarboxylate, 4-amino-1-carbethoxypiperidine, unii-54815srp4z, 1-piperidinecarboxylic acid, 4-amino-, ethyl ester, ethyl 4-aminopiperidinecarboxylate, ethyl-4-amino-1-piperidine carboxylate, 4-amino-1-piperidinecarboxylic acid ethyl ester, ethyl-4-aminopiperidin-1-carboxylat, pubchem2064, acmc-209m7k
PubChem CID 100867
IUPAC Name ethyl 4-aminopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)N

Specifications

Density 1.004
Boiling Point 118°C to 120°C (6 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.483
Quantity 25 g
Beilstein 388597
Sensitivity Air Sensitive
Formula Weight 172.23
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminopiperidine-1-carboxylate
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 261-472-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.