Learn More
Sigma Organic Chemistry Ethyl 4-aminotetrahydro | 1G | MFCD20526754
Supplier: Sigma Organic Chemistry CBR016701G

Ethyl 4-aminotetrahydro , 1G
About this Item:
MDL #: MFCD20526754
Molecular Weight: 209.67
UNSPSC Code: 12352200
Chemical Formula: C8H16ClNO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.