Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-bromobenzoate, 98+%

Catalog No. p-7081407
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol

Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5798-75-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016329
InChI Key XZIAFENWXIQIKR-UHFFFAOYSA-N
Synonym 4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate
PubChem CID 22043
IUPAC Name ethyl 4-bromobenzoate
SMILES CCOC(=O)C1=CC=C(Br)C=C1

Specifications

Density 1.403
Boiling Point 262°C to 264°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 10 g
Beilstein 1867129
Merck Index 14,1408
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-bromobenzoate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-347-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.