Alfa Aesar™ Ethyl 4-bromobenzoate, 98+%

Manufacturer: Alfa Aesar™ A1136509

Quantity 10g

 View more versions of this product

Catalog No. AAA1136509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5798-75-4
    Molecular Formula C9H9BrO2
    Formula Weight 229.07
    MDL Number MFCD00016329
    Beilstein 1867129
    Merck Index 14,1408
    Synonym 4-Bromobenzoic acid ethyl ester
    Chemical Name or Material Ethyl 4-bromobenzoate

    Specifications

    Formula Weight 229.07
    TSCA TSCA
    Synonym 4-Bromobenzoic acid ethyl ester
    Molecular Formula C9H9BrO2
    Quantity 10g
    Flash Point >110°C (230°F)
    MDL Number MFCD00016329
    Boiling Point 262° to 264°C
    CAS 5798-75-4
    Beilstein 1867129
    Merck Index 14,1408
    Chemical Name or Material Ethyl 4-bromobenzoate
    Assay ≥98%

    Description

    Applications
    Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit