Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-bromobenzoate, 99%

Catalog No. p-127275
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle

CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5798-75-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016329
InChI Key XZIAFENWXIQIKR-UHFFFAOYSA-N
Synonym 4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate
PubChem CID 22043
IUPAC Name ethyl 4-bromobenzoate
SMILES CCOC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 18°C
Density 1.4300g/mL
Boiling Point 262°C to 264°C
Flash Point >110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5440 to 1.547
Linear Formula BrC6H4CO2CH2CH3
Quantity 10 mL
Specific Gravity 1.43
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
Formula Weight 229.06
Percent Purity 99%
Chemical Name or Material Ethyl 4-bromobenzoate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 227-347-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.