Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-bromobutyrate, 98%

Catalog No. AAA134700B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA134700B 1000 g
AAA1347018 50 g
AAA1347030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA134700B Supplier Thermo Scientific Chemicals Supplier No. A134700B
Add to Cart

CAS: 2969-81-5 | C6H11BrO2 | 195.056 g/mol

Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

Solubility
Immiscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000259
InChI Key XBPOBCXHALHJFP-UHFFFAOYSA-N
Synonym ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
PubChem CID 76300
IUPAC Name ethyl 4-bromobutanoate
SMILES CCOC(=O)CCCBr

Specifications

Density 1.363
Boiling Point 80°C to 82°C (10 mmHg)
Flash Point 90°C (194°F)
Odor Odorless
Assay Percent Range 98%
Refractive Index 1.456
Quantity 1000 g
Beilstein 1749700
Solubility Information Immiscible with water.
Formula Weight 195.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromobutyrate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 221-005-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.