Alfa Aesar™ Ethyl 4-bromocrotonate, tech. 80%

Manufacturer: Alfa Aesar™ A1037109

Quantity 10g

 View more versions of this product

Catalog No. AAA1037109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37746-78-4
    Molecular Formula BrCH2CH=CHCO2CH2CH3
    Formula Weight 193.05
    MDL Number MFCD00000247
    UN Number UN1760
    Beilstein 1721388
    Synonym 4-Bromocrotonic acid ethyl ester
    Chemical Name or Material Ethyl 4-bromocrotonate
    Grade Technical Grade

    Specifications

    Formula Weight 193.05
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 37746-78-4
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Molecular Formula BrCH2CH=CHCO2CH2CH3
    Beilstein 1721388
    Quantity 10g
    Flash Point 97°C (206°F)
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00000247
    UN Number UN1760
    Synonym 4-Bromocrotonic acid ethyl ester
    Health Hazard 1 Danger
    Boiling Point 94° to 95°C (12mmHg)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Chemical Name or Material Ethyl 4-bromocrotonate
    Assay 80%
    Grade Technical Grade

    Description

    Applications
    Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

    Solubility
    Insoluble in water.

    Notes
    Store at 4°C. Incompatible with acids, bases, oxidizing agents, reducing agents. Protect from heat. Store away from strong bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2°C to 8°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit