Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chloro-7-methoxyquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7039948
Click to view available options
Quantity:
1 g

Catalog No. AAH5947303

Supplier: Thermo Scientific Chemicals H5947303

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77156-85-5
Molecular Formula C13H12ClNO3
Molecular Weight (g/mol) 265.693
MDL Number MFCD02217815
InChI Key IXMRUGGFGJIANG-UHFFFAOYSA-N
Synonym ethyl 4-chloro-7-methoxy-3-quinolinecarboxylate, 4-chloro-7-methoxyquinoline-3-carboxylic acid ethyl ester, cbmicro_031264, ethyl 4-chloro-7-methoxyquinoline-3-carboxylate, 4-chloro-7-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-3-ethoxycarbonyl-7-methoxyquinoline, 7-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4-chloro-7-methoxy-quinoline-3-carboxylate
PubChem CID 728976
IUPAC Name ethyl 4-chloro-7-methoxyquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=C(C=CC2=C1Cl)OC

Specifications

Quantity 1 g
Formula Weight 265.7
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-7-methoxyquinoline-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.