Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chloroquinoline-3-carboxylate, 96%, Thermo Scientific™

Catalog No. p-7033135
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-chloroquinoline-3-carboxylate is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13720-94-0
Molecular Formula C12H10ClNO2
Molecular Weight (g/mol) 235.667
MDL Number MFCD00173407
InChI Key DWXQUAHMZWZXHP-UHFFFAOYSA-N
Synonym ethyl 4-chloro-3-quinolinecarboxylate, 4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-quinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4-chloro-, ethyl ester, pubchem19592, acmc-20ai5d, ethylchloroquinolinecarboxylate, ethyl 4-chloroquinolin-3-carboxylate, ethyl 4-chloro-3-quinoline carboxylate, ethyl 4-chloranylquinoline-3-carboxylate
PubChem CID 268963
IUPAC Name ethyl 4-chloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC=C2N=C1)Cl

Specifications

Quantity 250 mg
Formula Weight 235.67
Percent Purity 96%
Chemical Name or Material Ethyl 4-chloroquinoline-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.