Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Cyanobenzoate 98.0+%, TCI America™
SDP

Catalog No. E116725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7153-22-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00001824
InChI Key JLSSWDFCYXSLQX-UHFFFAOYSA-N
Synonym benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester
PubChem CID 81589
IUPAC Name ethyl 4-cyanobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 55°C
Color White
Boiling Point 140°C
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Cyanobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.