Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-hydroxy-3-methoxycinnamate, 98%

Catalog No. AAL1496003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1496003 1 g
AAL14960MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1496003 Supplier Thermo Scientific Chemicals Supplier No. L1496003
Add to Cart

CAS: 4046-02-0 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4046-02-0
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009190
InChI Key ATJVZXXHKSYELS-FNORWQNLSA-N
Synonym ethyl ferulate, ferulic acid ethyl ester, ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, unii-5b8915uelw, ethyl 4'-hydroxy-3'-methoxycinnamate, e-ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, ethyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, ethyl 4-hydroxy-3-methoxycinnamate, e-3-4-hydroxy-3-methoxy-phenyl-acrylic acid ethyl ester, ethyl 4-hydroxy-3-methoxycinnamate ferulic acid ethyl ester
PubChem CID 736681
IUPAC Name ethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Beilstein 2696807
Sensitivity Light sensitive
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Ethyl 4-hydroxy-3-methoxycinnamate
EINECSNumber 223-745-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.