Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) nitrate, 99.5%

Catalog No. AC265590250
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 10102-45-1 | NO3Tl | 266.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-45-1
Molecular Formula NO3Tl
Molecular Weight (g/mol) 266.38
MDL Number MFCD00011280
InChI Key FYWSTUCDSVYLPV-UHFFFAOYSA-N
Synonym thallium i nitrate, thallium 1+ nitrate, thallous nitrate, thallium mononitrate, thallium nitrate van, thallium nitrate, tlno3, unii-ljq38dsr12, rcra waste number u217, thallium i nitrate 1:1
PubChem CID 24937
IUPAC Name thallium(1+);nitrate
SMILES [Tl+].[O-][N+]([O-])=O

Specifications

Boiling Point 433.0°C
Melting Point 206.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.5%
Linear Formula TlNO3
Fieser 13,86
Merck Index 15, 9414
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 266.38
Percent Purity 99.5%
Packaging Glass bottle
Chemical Name or Material Thallium(I) nitrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 233-273-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.