Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Oxo-3-piperidinecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. E089625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E089625G 25 g
E08965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E089625G Supplier TCI America Supplier No. E089625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4644-61-5
Molecular Formula C8H14ClNO3
Molecular Weight (g/mol) 207.654
MDL Number MFCD00043267
InChI Key RTYXWLMQWSFXNI-UHFFFAOYSA-N
Synonym ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
PubChem CID 107315
IUPAC Name ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point 169°C
Quantity 25 g
Formula Weight 207.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Oxo-3-piperidinecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.