Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. I0294100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0294100ML 100 mL
I029425ML 25 mL
I0294500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0294100ML Supplier TCI America Supplier No. I0294100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1126-09-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00006003
InChI Key RUJPPJYDHHAEEK-UHFFFAOYSA-N
Synonym ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
PubChem CID 70770
IUPAC Name ethyl piperidine-4-carboxylate
SMILES CCOC(=O)C1CCNCC1

Specifications

Color Yellow
Boiling Point 105°C
Quantity 100 mL
Formula Weight 157.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.