Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5,5-dimethyl-2,4-dioxohexanoate, 97%, Thermo Scientific™

Catalog No. p-4510606
Click to view available options
Quantity:
25 g

Catalog No. TL00361EB

Supplier: Thermo Scientific Chemicals TL00361EB

Only null left
Add to Cart

Chemical Identifiers

CAS 13395-36-3
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
InChI Key NIMKIMUBJFWPTD-UHFFFAOYSA-N
Synonym ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
PubChem CID 83403
IUPAC Name ethyl 5,5-dimethyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)(C)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Ethyl 5, 5-dimethyl-2, 4-dioxohexanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.