Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-bromonicotinate, 98%

Catalog No. p-7025354
Click to view available options
Quantity:
5 g
25 g

CAS: 20986-40-7 | C8H8BrNO2 | 230.061 g/mol

Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20986-40-7
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00040366
InChI Key PCPIANOJERKFJI-UHFFFAOYSA-N
Synonym ethyl 5-bromonicotinate, 5-bromonicotinic acid ethyl ester, ethyl 5-bromo-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 5-bromo-, ethyl ester, 5-bromo-3-pyridinecarboxylic acid ethyl ester, ethyl5-bromonicotinate, 5-bromo nicotinic acid ethyl ester, 5-bromo-nicotinic acid ethyl ester, ethyl 3-bromo-5-pyridinecarboxylate, 3-bromo-5-pyridine-carboxylic acid ethyl ester
PubChem CID 140785
IUPAC Name ethyl 5-bromopyridine-3-carboxylate
SMILES CCOC(=O)C1=CC(=CN=C1)Br

Specifications

Melting Point 39°C to 42°C
Boiling Point 86°C to 92°C (0.5 mmHg)
Quantity 5 g
Beilstein 131473
Formula Weight 230.07
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.