Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH6130403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6130403 1 g
AAH61304MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6130403 Supplier Thermo Scientific Chemicals Supplier No. H6130403
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197193-30-8
Molecular Formula C7H7BrN2O2
Molecular Weight (g/mol) 231.049
MDL Number MFCD12964050
InChI Key CZQFHUXIWZYQGP-UHFFFAOYSA-N
Synonym ethyl 5-bromopyrimidine-2-carboxyate, ethyl-5-bromopyrimidine-2-carboxylate, 5-bromo-2-pyrimidinecarboxylic acid ethyl ester, ethyl5-bromopyrimidine-2-carboxylate, 5-bromopyrimidine-2-carboxylic acid ethyl ester
PubChem CID 52987579
IUPAC Name ethyl 5-bromopyrimidine-2-carboxylate
SMILES CCOC(=O)C1=NC=C(C=N1)Br

Specifications

Color White
Quantity 1 g
Formula Weight 231.05
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Ethyl 5-bromopyrimidine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.