Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 5-chloropyrazine-2-carboxylate, 95%

Click to view available options
Quantity:
250mg
1g
54013-04-6
C7H7ClN2O2
186.595
MFCD18070850
CFKGDFQQELWOHA-UHFFFAOYSA-N
ethyl 5-chloropyrazinoate,pyrazinecarboxylic acid, 5-chloro-, ethyl ester,ethyl 5-chloro-2-pyrazinecarboxylate,ethyl5-chloropyrazine-2-carboxylate,5-chloro-2-pyrazinecarboxylic acid ethyl ester
465025
ethyl 5-chloropyrazine-2-carboxylate
CCOC(=O)C1=CN=C(C=N1)Cl
54013-04-6
C7H7ClN2O2
186.595
MFCD18070850
CFKGDFQQELWOHA-UHFFFAOYSA-N
ethyl 5-chloropyrazinoate,pyrazinecarboxylic acid, 5-chloro-, ethyl ester,ethyl 5-chloro-2-pyrazinecarboxylate,ethyl5-chloropyrazine-2-carboxylate,5-chloro-2-pyrazinecarboxylic acid ethyl ester
465025
ethyl 5-chloropyrazine-2-carboxylate
CCOC(=O)C1=CN=C(C=N1)Cl
Quantity 250mg
Beilstein 779320
Formula Weight 186.6
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Ethyl 5-chloropyrazine-2-carboxylate