Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 7-Bromoheptanoate 97.0+%, TCI America™
SDP

Catalog No. B188025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B188025G 25 g
B18805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B188025G Supplier TCI America Supplier No. B188025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29823-18-5
Molecular Formula C9H17BrO2
Molecular Weight (g/mol) 237.14
MDL Number MFCD00037103
InChI Key OOBFNDGMAGSNKA-UHFFFAOYSA-N
PubChem CID 122490
IUPAC Name ethyl 7-bromoheptanoate
SMILES CCOC(=O)CCCCCCBr

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
Synonym 7-bromoheptanoic acid ethyl ester, ethyl-7-bromoheptanoate, heptanoic acid, 7-bromo-, ethyl ester, ethyl7-bromoheptanoate, br ch2 6cooet, ethyl 7-bromo-heptanoate, ethyl 7-bromanylheptanoate, acmc-209hb9, ethyl 7-bromoheptanoate, oobfndgmagsnka-uhfffaoysa
Formula Weight 237.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 7-Bromoheptanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.