Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, EMPLURA™, MilliporeSigma™

Catalog No. m8222775000
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83°C
Density 0.90g/cm3
Boiling Point 777 deg;C (at 1013 hPa)
Quantity 5 L
Explosion Limit 2.1 - 11.5%(V)
Flash Point -4°C
Ignition Point 460°C
Packaging Plastic Bottle
Assay Percent Range ≥99.5% (a/a)
Residue after Evaporation ≤0.003%
Linear Formula C4H8O2
Vapor Pressure 97 hPa
UN Number UN 1173
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Category Danger
Hazard Statement H225: Highly flammable liquid and vapour. H319: Causes serious eye irritation. H336: May cause drowsiness or dizziness. EUH066: Repeated exposure may cause skin dryness or cracking.
Precautionary Statement P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking. P240: Ground/bond container and receiving equipment. P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 205-500-4
RTECSNumber AH5425000
Recommended Storage Store at +2°C to +25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.