Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, 5% v/v Aqueous, Reagents

Catalog No. 01335311
Change view
Click to view available options
:
Quantity: 50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-311 Quantity: 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-311 Supplier Reagents Holdings Llc Supplier No. CE28000050C
Only null left
Add to Cart
Add to Cart

Ethyl Acetate, 5% v/v Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 141-78-6 , 7732-18-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Color Colorless
Quantity 50 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9948
Percent Purity 5%
Grade Reagent
Chemical Name or Material Ethyl Acetate, 5% v/v Aqueous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.