Learn More
Ethyl acetate, ACS, 99.5+%
Description
Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -84°C |
| Color | Colorless |
| Density | 0.902 g/mL |
| Boiling Point | 77°C |
| Quantity | 1 L |
| Flash Point | -3°C (26°F) |
| Odor | Sweet |
| Physical Form | Liquid |
| Refractive Index | 1.372 |
| Linear Formula | CH3CO2CH2CH3 |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H336 |
| Hazard Statement | GHS H Statement H225-H319-H336 Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE |
| EINECSNumber | 205-500-4 |
| RTECSNumber | AH5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.