Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, B&J Brand™, for HPLC, for spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. 6000583
Click to view available options
Quantity:
56 L
200 L
Packaging:
Returnable Steel Drum

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Absorbance 0.05 AU max. at 275 nm, 0.03 AU max. at 300 nm, 0.005 AU max. at 325 nm, 0.005 AU max. at 350 nm
CAS Max % 100.0000%
Density 0.8945 g/cm3
Boiling Point 77°C
Quantity 200 L
Flash Point −4°C
Packaging Returnable Steel Drum
UN Number UN1173
Beilstein 506104
Water 0.03% max.
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Ethyl Acetate, B&J Brand™, for HPLC, for spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.