Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Acetate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Specifications
Specifications
| Quantity | 1 L |
| Flash Point | −4.00°C |
| Packaging | Aluminum Bottle |
| Specific Gravity | 1L = 0.90kg |
| Formula Weight | 88.11 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Ethyl Acetate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction