Learn More
Ethyl Acetate (Certified ACS), Fisher Chemical PROMO
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | -83.5°C |
| Color | Colorless |
| Density | 0.902 g/cm3 |
| Boiling Point | 75°C |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | ≥99.5% |
| Residue after Evaporation | 0.003% Max |
| Show More |
Safety and Handling
| Hazard Category | DANGER! |
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview: Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.