Learn More
Ethyl acetate, HPLC Grade, 99.5+%
Description
Ethyl acetate in its highly pure form is used as a solvent for spectrophotometric applications. It is most suited for HPLC instruments and spectrophotometers including UV-visible spectrophotometer, IR spectrometer and mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. Also, it is used in glues, nail polish removers, decaffeinating tea and coffee, and cigarettes. It is present in confectioneries, perfumes, and fruits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 84°C |
| Color | Colorless |
| Appearance | Colorless liquid |
| Density | 0.902 g/cm3 |
| Boiling Point | 77°C |
| Quantity | 1 L |
| Flash Point | -3°C (26°F) |
| Ignition Point | 460°C (860°F) |
| Odor | Fruit-like |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H336 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3, Packing Group: II |
| EINECSNumber | 205-500-4 |
| RTECSNumber | AH5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.