Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Innovating Science™
SCI_ED

Catalog No. S031142
Click to view available options
Quantity:
30 mL
500 mL

Chemical Identifiers

Molecular Weight (g/mol) 88.11 g/mol

Specifications

Boiling Point 77°C
Quantity 30 mL
Packaging Amber Glass with Dropper
Physical Form Liquid
Percent Purity 99.5%
Chemical Name or Material Ethyl Acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.