Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Optima™ for HPLC and GC, Thermo Scientific™
GSA_VA

Catalog No. e196sk4 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 

Catalog No. E196SK-4

Only null left
Add to Cart

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 0.015 AU max. at 270nm, 0.12 AU max. at 260nm, 1.00 AU max. at 255nm
Color Colorless
Density 0.902g/cm³
Boiling Point 75°C
Quantity 4 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Assay Percent Range Assay: 99.9% min by GC
Residue after Evaporation 1ppm max.
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOC2H5
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
Water 0.02% max.
Formula Weight 88.11
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.