Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O

E1025, 141-78-6, C4H8O2

Spectrum™ Ethyl Acetate, Reagent, ACS , is a colorless liquid that smells like pear drops. Abbreviated as EA, it can be found in glues, nail polish removers, cigarettes and other products. Primarily used as a diluent because of its low cost and toxicity, it is also used in laboratory settings for column chromatography and extractions. Prone to hydrolysis and transesterification, it is rarely used as a solvent. As an ACS grade Reagent, Spectrum Chemical manufactured Ethyl Acetate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
CAS Max % 1
UN Number UN1173
Percent Purity 99.5%
Packaging Amber Glass Bottle
Residue after Evaporation 3e-005
Water 0.002
Quantity 500 mL