Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate (Sequencing), Fisher BioReagents

Catalog No. p-25739 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
This solvent is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 1 max. at 255nm, 0.15 max. at 260nm, 0.025 max. at 270nm
Color Colorless
Density 0.902g/cm³
Boiling Point 75°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 10ppm max.
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOCH2CH3
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
UN Number 1173
Water 0.02% max.
Merck Index 15, 3811
Formula Weight 88.11
Percent Purity ≥99.9%
Grade Sequencing
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.