Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl butyrylacetate, 98%

Catalog No. AC118191000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 3249-68-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3249-68-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009401
InChI Key KQWWVLVLVYYYDT-UHFFFAOYSA-N
Synonym ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
PubChem CID 238498
ChEBI CHEBI:18119
IUPAC Name ethyl 3-oxohexanoate
SMILES CCCC(=O)CC(=O)OCC

Specifications

Color Colorless
Density 1.0000g/mL
Boiling Point 104.0°C (22.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4265 to 1.4285
Linear Formula CH3CH2CH2COCH2CO2C2H5
Quantity 100 g
Beilstein 03, 684
Specific Gravity 1
Solubility Information Solubility in water: immiscible with water
Formula Weight 158.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl butyrylacetate, 98%
Show More Show Less
EINECSNumber 221-835-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.