Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetoacetate, 99+%

Catalog No. AAA125440E
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g

CAS: 141-97-9 | C6H10O3 | 130.143 g/mol

Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

Solubility
Miscible with water, benzene, chloroform, ether and acetone.

Notes
Incompatible with acids, bases, oxidizing agents, alkali metals and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-97-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00009199
InChI Key XYIBRDXRRQCHLP-UHFFFAOYSA-N
Synonym ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
PubChem CID 8868
ChEBI CHEBI:4893
IUPAC Name ethyl 3-oxobutanoate
SMILES CCOC(=O)CC(=O)C

Specifications

Melting Point -43°C
Density 1.025
Boiling Point 180°C to 181°C
Flash Point 84°C (183°F)
Assay Percent Range ≥99%
Refractive Index 1.419
Quantity 2500 g
Beilstein 385838
Merck Index 14,3758
Solubility Information Miscible with water,benzene,chloroform,ether and acetone.
Formula Weight 130.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Ethyl Acetoacetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H319-H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 205-516-1
RTECSNumber AK5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.