Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetohydroximate 95.0+%, TCI America™
SDP

Catalog No. A133925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A133925G 25 g
A13395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A133925G Supplier TCI America Supplier No. A133925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10576-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00002114,MFCD00002114
InChI Key QWKAVVNRCKPKNM-SNAWJCMRSA-N
Synonym ethyl n-hydroxyacetimidate, n-hydroxyethanimidic acid, ethyl ester, ethyl acetohydroximate, ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyacetimidate, ethyl 1e-n-hydroxyethanimidate, snx`lfelmmpqh, ethyl acethydroxamate, 1-ethoxyethanone oxime
PubChem CID 6386647
IUPAC Name (E)-(ethyl N-hydroxyethanimidate)
SMILES CCO\C(C)=N\O

Specifications

Color White-Yellow
Boiling Point 60°C
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl Acetohydroximate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.