Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol

Catalog No. aaa15493ae
Click to view available options
Quantity:
100 mL
500 mL
2 L

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol

Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-88-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009188
InChI Key JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Synonym ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
PubChem CID 8821
ChEBI CHEBI:82327
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifications

Melting Point -71°C
Density 0.924
Boiling Point 99°C to 100°C
Flash Point −1°C (30°F)
Odor Pungent
Refractive Index 1.406
Linear Formula CH2=CHCO2CH2CH3
Quantity 100 mL
UN Number UN1917
Beilstein 773866
Merck Index 14,3759
Solubility Information Miscible with ethanol,ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Ethyl acrylate, stabilized with ≈20ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H302+H312-H315-H317-H319-H331-H335-H351
Hazard Statement GHS H Statement
H225-H302-H312-H332-H315-H319-H317-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P333+P313-P363
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACRYLATE, STABILIZED
EINECSNumber 205-438-8
RTECSNumber AT0700000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.