Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol Standard, 17% (v/v), Ricca Chemical
SDP

Catalog No. p-3148436 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Packaging Natural Poly Bottle
CAS Min % 82.0, 16.0
CAS Max % 84.0, 18.0
Melting Point Approximately 0°C
Type Standard
Quantity 50 mL
Concentration or Composition (by Analyte or Components) 17% (v/v)
Chemical Name or Material Ethyl Alcohol Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.