Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 190 Proof ACS USP, Greenfield

Catalog No. 502946028
Click to view available options
Packaging:
4x4L Case
Metal Drum
Poly Pail

Chemical Identifiers

CAS 64-17-5 (Ethyl Alcohol)
Molecular Weight (g/mol) 46.07 (Ethyl Alcohol)

Specifications

Boiling Point > 172.4 - < 176 °F (> 78 - < 80 °C)
Quantity 5 Gallon
Packaging Poly Pail
Percent Purity 95% (190 Proof)
Grade ACS/NF/USP
Chemical Name or Material Ethyl Alcohol 95% (190 Proof)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.