Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 200 Proof, 99.5% min., ACS, MilliporeSigma™

Catalog No. p-4382110
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Glass Bottle
Metal Drum

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Density 0.79
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.5% min.
Vapor Pressure 5.9 kPa (44mm Hg) [20°C]
Formula Weight 46.07
Chemical Name or Material Ethyl Alcohol, 200 Proof, 200 Proof, For ACS analysis
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.