Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18602522 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum

Catalog No. 18-602-522

Supplier: Spectrum Chemical Manufacturing Corporation E1028100ML

E1028, 64-17-5, C2H5OH

Spectrum™ Chemical Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS is a pure, 99.5% minimum assay, undenatured ethyl alcohol (ethanol) that is ideal for a variety of research, industrial and educational uses. Spectrum certifies that the Reagent ACS grade is perfect for general laboratory use and offers a solvent that will yield repetitive, consistent results.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Boiling Point 78°C to 79°C
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
Titratable Acid 0.0005 meq/g
Titratable Base 0.0002 meq/g
UN Number UN1170
Concentration 100%
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Ethyl Alcohol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.