Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Alcohol, Lab Grade, 95% Denatured w/IPA and Methanol, Fisher Science Education™ PROMO
Shop All Fisherbrand Products

Click to view available options
Quantity:
4 L
Specifications
Specifications
Color | Colorless |
Density | Approx. 0.8 g/mL |
Quantity | 4 L |
Packaging | Lab Grade - 95% Denatured w/IPA and Methanol |
Physical Form | Liquid |
Linear Formula | C2H5OH |
Formula Weight | 46.07 g/mol |
Chemical Name or Material | Ethyl Alcohol |
Safety and Handling

ShelfLife | 5 years |
DOTInformation | DOT Class 3, II, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction