Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ethyl Alcohol, Lab Grade, 95% Denatured w/IPA and Methanol
SCI_ED

Catalog No. s25309 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
3.8 L
4 L
Packaging:
Lab Grade - 95% Denatured w/IPA and Methanol
Lab Grade - 95% Denatured w/IPA and Methanol; Safecote™

Versatile 95% denatured ethyl alcohol ideal for science labs and instructional use.

  • Intended for instructional and demonstration use in academic settings
  • Ethyl Alcohol (Denatured)
  • C2H5OH ∣ Formula Weight: 46.07
  • 95% denatured with isopropyl alcohol and methanol
  • Liquid form
  • Approximately 0.8 g/mL
  • 1 liter
  • Lab-grade liquid chemical container
  • For educational use only – Certificate of Analysis (C of A) not available

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Density ∽0.8g/mL
Quantity 500 mL
Packaging Lab Grade - 95% Denatured w/IPA and Methanol
Physical Form Liquid
Linear Formula C2H5OH
Formula Weight 46.07
Chemical Name or Material Ethyl Alcohol
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.