Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl benzoate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187776 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Ethyl benzoate belongs to the class of aromatic esters.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 914°F
CAS 93-89-0
Vapor Pressure 1 mmHg (44°C)
Vapor Density 5.17 (vs. air)
Density 1.045 g/mL (at 25°C)
Flash Point 88°C
Physical Form Neat
Linear Formula C6H5 COOC2H5
MDL Number MFCD00009109
Melting Point -34°C
Boiling Point 212°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 150.17
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.