Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Bromofluoroacetate 98.0+%, TCI America™
SDP

Catalog No. B167325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 401-55-8
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.992
MDL Number MFCD00042095
InChI Key ULNDTPIRBQGESN-UHFFFAOYSA-N
Synonym ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t
PubChem CID 2733407
IUPAC Name ethyl 2-bromo-2-fluoroacetate
SMILES CCOC(=O)C(F)Br

Specifications

Color Yellow
Boiling Point 68°C
Quantity 25 g
UN Number 2810
Formula Weight 184.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Bromofluoroacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.