Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Butyrate 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
105-54-4
C6H12O2
116.16
MFCD00009394
OBNCKNCVKJNDBV-UHFFFAOYSA-N
ethyl butyrate,butyric acid ethyl ester,ethyl n-butyrate,butanoic acid, ethyl ester,butyric ester,butyric ether,butyric acid, ethyl ester,ethyl n-butanoate,butanoic acid ethyl ester,fema number 2427
7762
ethyl butanoate
CCCC(=O)OCC
105-54-4
C6H12O2
116.16
MFCD00009394
OBNCKNCVKJNDBV-UHFFFAOYSA-N
ethyl butyrate,butyric acid ethyl ester,ethyl n-butyrate,butanoic acid, ethyl ester,butyric ester,butyric ether,butyric acid, ethyl ester,ethyl n-butanoate,butanoic acid ethyl ester,fema number 2427
7762
ethyl butanoate
CCCC(=O)OCC
Packaging Glass bottle
Flash Point 26°C
Density 0.8700g/mL
Acidity 1mg KOH/g max.
Boiling Point 120.0°C
Color Colorless
Quantity 5mL
Melting Point -93.3°C
Infrared Spectrum Authentic
Refractive Index 1.3910 to 1.3930
Specific Gravity 0.87
Water 0.3% max. (K.F.)
Assay Percent Range 99%
Linear Formula CH3CH2CH2CO2C2H5
Beilstein 02, 270
Merck Index 15, 3830
Solubility Information Solubility in water: practically insoluble
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethyl butyrate, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.

GHS P Statement:
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-306-4

RTECSNumber : ET1660000

TSCA : TSCA