Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Butyrylacetate 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3249-68-1
C8H14O3
158.2
MFCD00009401
KQWWVLVLVYYYDT-UHFFFAOYSA-N
ethyl butyrylacetate,ethyl butyroacetate,hexanoic acid, 3-oxo-, ethyl ester,ethyl beta-ketohexanoate,ethyl alpha-butyrylacetate,3-ketohexanoic acid ethyl ester,ethyl-3-oxohexanoate,unii-8q1ahg710e,ethyl .alpha.-butyrylacetate,fema no. 3683
238498
CHEBI:18119
ethyl 3-oxohexanoate
CCCC(=O)CC(=O)OCC
3249-68-1
C8H14O3
158.2
MFCD00009401
KQWWVLVLVYYYDT-UHFFFAOYSA-N
ethyl butyrylacetate,ethyl butyroacetate,hexanoic acid, 3-oxo-, ethyl ester,ethyl beta-ketohexanoate,ethyl alpha-butyrylacetate,3-ketohexanoic acid ethyl ester,ethyl-3-oxohexanoate,unii-8q1ahg710e,ethyl .alpha.-butyrylacetate,fema no. 3683
238498
CHEBI:18119
ethyl 3-oxohexanoate
CCCC(=O)CC(=O)OCC
Color Colorless
Flash Point 78°C
Quantity 25g
Density 1.0000g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4265 to 1.4285
Boiling Point 104.0°C (22.0 mmHg)
Specific Gravity 1
Assay Percent Range 98%
Linear Formula CH3CH2CH2COCH2CO2C2H5
Beilstein 03, 684
Solubility Information Solubility in water: immiscible with water
Formula Weight 158.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl butyrylacetate, 98%
Show More Show Less

EINECSNumber : 221-835-9