Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl chloroformate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
Melting Point | -81.0°C |
Density | 1.1350g/mL |
Boiling Point | 94.0°C |
Flash Point | 10°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 1% max. (diethyl carbonate) (GC) |
Packaging | Glass bottle |
Molecular Formula | C3H5ClO2 |
Refractive Index | 1.3950 to 1.3960 |
Linear Formula | ClCO2C2H5 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Fatal if inhaled. Causes severe skin burns and eye damage. Toxic if swallowed. Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physi |
EINECSNumber | 208-778-5 |
RTECSNumber | LQ6125000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction